Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
axo
Entrez Gene ID
43923
Gene Name
axotactin
Gene Aliases
CG13715, CG13717, CG18296, CG32234, CG42664, CG43225, CT1277, Dmel\CG43225, Dmel_CG43225
Chromosome Location
Chr.3L: 4629604 - 4687317 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0262870
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001259701.1 NP_001246630.1 2 - 3 801 68
NM_001259702.1 NP_001246631.1 2 - 3 801 68
NM_001259703.1 NP_001246632.1 2 - 3 801 68
NM_001274493.1 NP_001261422.1 2 - 3 910 68
NM_001274494.1 NP_001261423.1 2 - 3 910 68
NM_001274495.1 NP_001261424.1 2 - 3 910 68
NM_079917.5 NP_524656.5 2 - 3 910 68

View Full Product Details