Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- prc
- Entrez Gene ID
-
43930
- Gene Name
- pericardin
- Gene Aliases
-
BEST:CK01593, CG5700, CK01593, Dmel\CG5700, Dmel_CG5700, EC11, PC, PRC, Pc, Prc, pc
- Chromosome Location
-
Chr.3L: 11958872 - 11966131 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0028573
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079919.3 |
NP_524658.2 |
5 - 6 |
546 |
63 |
View Full Product Details