Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cds
Entrez Gene ID
43950
Gene Name
CDP-diacylglycerol synthase
Gene Aliases
CDS, CG7962, CdsA, Dmel\CG7962, Dmel_CG7962, cds, cdsA, eye-CDS, eye-cds
Chromosome Location
Chr.3L: 8127762 - 8131675 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0010350
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300047.1 NP_001286976.1 2 - 3 533 78
NM_079922.4 NP_524661.1 1 - 2 315 78

View Full Product Details