Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
spag
Entrez Gene ID
43958
Gene Name
spaghetti
Gene Aliases
CG13570, Dmel\CG13570, Dmel_CG13570, Rpap3, SPAG, Spag, l(2)k12101, spagh
Chromosome Location
Chr.2R: 24159384 - 24161503 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0015544
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079925.3 NP_524664.1 1 - 2 239 77
GenBank mRNA AJ252086.2 - 1 - 2 258 77
BT150413.1 - 1 - 2 235 77

View Full Product Details