Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Plap
Entrez Gene ID
43978
Gene Name
Phospholipase A2 activator protein
Gene Aliases
CG5105, Dmel\CG5105, Dmel_CG5105, UFD3, dPLAP, dmPLAP, plap
Chromosome Location
Chr.2L: 1167172 - 1170365 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0024314
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001258900.2 NP_001245829.1 3 - 4 1811 70
NM_001298628.1 NP_001285557.1 4 - 5 1965 70
NM_079927.3 NP_524666.2 3 - 4 1948 70
GenBank mRNA AY058773.1 - 3 - 4 1779 70

View Full Product Details