Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Optix
Entrez Gene ID
44108
Gene Name
CG18455-PA
Gene Aliases
CG18455, D-Six3, Dmel\CG18455, Dmel_CG18455, Dsix3, OPTIX, Six3, anon-WO0153538.79, opt, optix, opx, six3
Chromosome Location
Chr.2R: 8031283 - 8042529 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0025360
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273864.2 NP_001260793.1 3 - 4 1328 86
NM_079956.3 NP_524695.2 3 - 4 1018 86
GenBank mRNA AY061077.1 - 3 - 4 978 86

View Full Product Details