Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sut3
Entrez Gene ID
44268
Gene Name
sugar transporter 3
Gene Aliases
CG17976, Dmel\CG17976, Dmel_CG17976
Chromosome Location
Chr.2R: 8089482 - 8091297 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0028561
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273869.1 NP_001260798.1 3 - 4 1117 88
NM_079992.4 NP_524731.1 3 - 4 1125 88
GenBank mRNA AF199485.1 - 3 - 4 1097 88
BT044406.1 - 3 - 4 1125 88

View Full Product Details