Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tio
Entrez Gene ID
44272
Gene Name
tiptop
Gene Aliases
CG12630, Dmel\CG12630, Dmel_CG12630, Tio
Chromosome Location
Chr.2L: 22019208 - 22025734 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0028979
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273750.1 NP_001260679.1 2 - 3 3338 61
NM_079994.4 NP_524733.2 2 - 3 3224 61

View Full Product Details