Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
arr
Entrez Gene ID
44279
Gene Name
arrow
Gene Aliases
Arr, BEST:CK00539, CG5912, CK00539, CT15575, Dm Arr, Dmel\CG5912, Dmel_CG5912, Irp6, LRP, LRP/Arr, LRP5/6, LRP6, l(2)k08131
Chromosome Location
Chr.2R: 13453640 - 13483229 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0000119
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079998.4 NP_524737.2 3 - 4 3376 88
GenBank mRNA BT133102.1 - 3 - 4 3359 88

View Full Product Details