Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- arr
- Entrez Gene ID
-
44279
- Gene Name
- arrow
- Gene Aliases
-
Arr, BEST:CK00539, CG5912, CK00539, CT15575, Dm Arr, Dmel\CG5912, Dmel_CG5912, Irp6, LRP, LRP/Arr, LRP5/6, LRP6, l(2)k08131
- Chromosome Location
-
Chr.2R: 13453640 - 13483229 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0000119
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079998.4 |
NP_524737.2 |
5 - 6 |
3801 |
73 |
GenBank mRNA |
BT133102.1 |
- |
5 - 6 |
3784 |
73 |
View Full Product Details