Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Ptp52F
- Entrez Gene ID
-
36790
- Gene Name
- Protein tyrosine phosphatase 52F
- Gene Aliases
-
CG18243, CT18044, DPTP52F, Dmel\CG18243, Dmel_CG18243, PTP52F, ptp52F
- Chromosome Location
-
Chr.2R: 16171112 - 16178307 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0034085
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_137249.4 |
NP_611093.2 |
15 - 16 |
3636 |
63 |
GenBank mRNA |
BT133387.1 |
- |
15 - 16 |
3616 |
63 |
View Full Product Details