Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- rdgBbeta
- Entrez Gene ID
-
37011
- Gene Name
- CG17818-PA
- Gene Aliases
-
BcDNA.LD14189, BcDNA:LD14189, CG17818, Dmel\CG17818, Dmel_CG17818, DrdgBbeta, RdgB-beta, RdgBbeta
- Chromosome Location
-
Chr.2R: 17695776 - 17698362 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0027872
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_137404.5 |
NP_611248.3 |
6 - 7 |
605 |
93 |
GenBank mRNA |
AF160934.1 |
- |
6 - 7 |
571 |
93 |
View Full Product Details