Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- GstE5
- Entrez Gene ID
-
37110
- Gene Name
- Glutathione S transferase E5
- Gene Aliases
-
CG17527, DmGSTE5, Dmel\CG17527, Dmel_CG17527, GSTE5, gstE5, gste5
- Chromosome Location
-
Chr.2R: 18404605 - 18405338 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0063495
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_137483.3 |
NP_611327.1 |
1 - 1 |
651 |
78 |
View Full Product Details