Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- isopeptidase-T-3
- Entrez Gene ID
-
37261
- Gene Name
- CG11025-PA
- Gene Aliases
-
CG11025, Dmel\CG11025, Dmel_CG11025, ISOT-3, ISOT-3A, IsoT-3, Isopeptidase-T-3
- Chromosome Location
-
Chr.2R: 19670618 - 19673477 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0028372
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_137595.2 |
NP_611439.2 |
1 - 2 |
528 |
60 |
GenBank mRNA |
BT004899.1 |
- |
1 - 2 |
435 |
60 |
View Full Product Details