Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- rig
- Entrez Gene ID
-
37335
- Gene Name
- rigor mortis
- Gene Aliases
-
CG11171, CG13440, CG30149, Dmel\CG30149, Dmel_CG30149, Gem5, Rig, anon-SAGE:Wang-132, l(2)05056
- Chromosome Location
-
Chr.2R: 20633876 - 20640681 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0250850
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_137655.3 |
NP_611499.3 |
6 - 7 |
3225 |
62 |
GenBank mRNA |
AY274835.1 |
- |
6 - 7 |
3210 |
62 |
View Full Product Details