Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ipk1
Entrez Gene ID
37360
Gene Name
CG30295-PA
Gene Aliases
CG15228, CG15229, CG15230, CG30295, Dmel\CG30295, Dmel_CG30295, dmIpk1
Chromosome Location
Chr.2R: 20870791 - 20879145 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0050295
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103917.3 NP_001097387.1 4 - 5 1057 66
NM_001274182.2 NP_001261111.1 4 - 5 1057 66
NM_137678.3 NP_611522.1 4 - 5 1057 66
GenBank mRNA AY061821.1 - 4 - 5 1042 66

View Full Product Details