Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- cpa
- Entrez Gene ID
-
37387
- Gene Name
- capping protein alpha
- Gene Aliases
-
CG10540, Cap1, CapZalpha, Cpa, Dmel\CG10540, Dmel_CG10540, cpalpha, miro, scrd
- Chromosome Location
-
Chr.2R: 21057787 - 21059883 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0034577
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_137695.4 |
NP_611539.1 |
2 - 3 |
644 |
60 |
GenBank mRNA |
AY069084.1 |
- |
2 - 3 |
622 |
60 |
View Full Product Details