Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Cht9
- Entrez Gene ID
-
37392
- Gene Name
- Chitinase 9
- Gene Aliases
-
CG10531, DmCHT9, DmCht9, Dmel\CG10531, Dmel_CG10531
- Chromosome Location
-
Chr.2R: 21067992 - 21069286 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0034582
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_137699.5 |
NP_611543.3 |
1 - 2 |
71 |
57 |
View Full Product Details