Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Tpst
- Entrez Gene ID
-
32312
- Gene Name
- Tyrosylprotein sulfotransferase
- Gene Aliases
-
CG1573, CG15758, CG15759, CG32629, CG32629/CG32632, CG32632, DmTPST, Dmel\CG32632, Dmel_CG32632, TANGO13, TPST, Tango13
- Chromosome Location
-
Chr.X: 13563823 - 13593224 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0086674
- Gene Symbol
- CG46311
- Entrez Gene ID
-
30522400
- Gene Name
- CG46311-PA
- Gene Aliases
-
Dmel\CG46311, Dmel_CG46311, Tango13-5'utr
- Chromosome Location
-
Chr.X: 13563823 - 13590570 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0284227
View Full Product Details