Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pis
Entrez Gene ID
32506
Gene Name
Phosphatidylinositol synthase
Gene Aliases
CG9245, Dmel\CG9245, Dmel_CG9245, PIS, anon-WO02059370.38, dPIS, dPis, dpis
Chromosome Location
Chr.X: 15701451 - 15703770 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0030670
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_132827.3 NP_573055.1 1 - 2 200 70
NM_167456.3 NP_727878.1 1 - 2 200 70
GenBank mRNA AY094866.1 - 1 - 2 196 70

View Full Product Details