Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
Muc14A
Entrez Gene ID
318097
Gene Name
Mucin 14A
Gene Aliases
CG32580, Dmel\CG32580, Dmel_CG32580
Chromosome Location
Chr.X: 16009114 - 16061722 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0052580
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_167482.4 NP_727928.3 - 41571 123

View Full Product Details