Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
disco-r
Entrez Gene ID
64875
Gene Name
disco-related
Gene Aliases
CG32577, CG9219, CG9223, Dmel\CG32577, Dmel_CG32577, bae
Chromosome Location
Chr.X: 16118067 - 16148977 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0285879
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001272669.1 NP_001259598.1 3 - 4 2446 145
NM_001272670.2 NP_001259599.1 3 - 4 4276 145
NM_001272671.1 NP_001259600.1 2 - 3 2210 145
NM_167487.3 NP_727938.1 4 - 5 2744 145

View Full Product Details