Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- nonA
- Entrez Gene ID
-
32603
- Gene Name
- no on or off transient A
- Gene Aliases
-
Bj6, Bj6-1, CG4211, Dmel\CG4211, Dmel_CG4211, NONA, NonA, dNONO, diss, non-A, nonA/diss, opm2, p54[nrb], x14
- Chromosome Location
-
Chr.X: 16361569 - 16371547 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0004227
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_167505.2 |
NP_727962.1 |
4 - 5 |
2121 |
138 |
View Full Product Details