Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ankle2
Entrez Gene ID
32732
Gene Name
Ankyrin repeat and LEM domain containing 2
Gene Aliases
CG8465, Dmel\CG8465, Dmel_CG8465, anon-WO03040301.175, anon-WO03040301.177, anon-WO03040301.179, dAnkle2, l(1)G0222, l(1)G0316
Chromosome Location
Chr.X: 17571854 - 17579229 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0028343
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001298446.1 NP_001285375.1 4 - 5 1286 70
NM_132993.3 NP_573221.2 4 - 5 1286 70
NM_167569.2 NP_728082.2 3 - 4 1292 70
NM_167570.2 NP_728083.2 2 - 3 985 70
GenBank mRNA AY122241.1 - 3 - 4 1292 70

View Full Product Details