Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HisRS
Entrez Gene ID
32841
Gene Name
Histidyl-tRNA synthetase
Gene Aliases
Aats-his, CG6335, Dmel\CG6335, Dmel_CG6335, HRS, Harsl, dHARS, l(1)G0165, l(1)G0358
Chromosome Location
Chr.X: 18515428 - 18520445 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0027087
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001272756.1 NP_001259685.1 5 - 6 1791 99
NM_001272757.1 NP_001259686.1 5 - 6 2403 99
NM_133077.2 NP_573305.2 5 - 6 1645 99
NM_167624.3 NP_728180.1 4 - 5 1519 99
GenBank mRNA AY102665.1 - 4 - 5 1548 99

View Full Product Details