Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vav
Entrez Gene ID
32920
Gene Name
Vav guanine nucleotide exchange factor
Gene Aliases
BcDNA:GH01128, CG7893, DmVav, Dmel\CG7893, Dmel_CG7893, DroVav, vav
Chromosome Location
Chr.X: 19261820 - 19273280 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0040068
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103560.3 NP_001097030.1 2 - 3 953 58
NM_001272774.1 NP_001259703.1 2 - 3 953 58
NM_133144.4 NP_573372.1 2 - 3 953 58
NM_167642.3 NP_728235.1 2 - 3 953 58
GenBank mRNA AY069536.1 - 2 - 3 942 58

View Full Product Details