Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tao
Entrez Gene ID
32948
Gene Name
CG14217-PA
Gene Aliases
AAF48973, CG14217, Dmel\CG14217, Dmel_CG14217, MARKK, TAO1, Tao-1, Tao1, dTao-1, tao, tao-1, tao1
Chromosome Location
Chr.X: 19569721 - 19579800 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0031030
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001201743.3 NP_001188672.1 11 - 12 3386 88
NM_001201744.2 NP_001188673.1 7 - 8 1450 88
NM_001272779.1 NP_001259708.1 11 - 12 3581 88
NM_134475.4 NP_608319.1 7 - 8 1450 88
NM_167665.2 NP_728267.1 11 - 12 3036 88
NM_167666.3 NP_728268.1 11 - 12 3386 88
NM_167667.3 NP_728269.1 6 - 7 1589 88
GenBank mRNA AB277547.1 - 11 - 12 3365 88
AB277548.1 - 7 - 8 1376 88
AY069469.1 - 7 - 8 1419 88
BT099719.1 - 11 - 12 3325 88

View Full Product Details