Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tao
Entrez Gene ID
32948
Gene Name
CG14217-PA
Gene Aliases
AAF48973, CG14217, Dmel\CG14217, Dmel_CG14217, MARKK, TAO1, Tao-1, Tao1, dTao-1, tao, tao-1, tao1
Chromosome Location
Chr.X: 19569721 - 19579800 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0031030
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001201743.3 NP_001188672.1 6 - 7 2384 73
NM_001201744.2 NP_001188673.1 2 - 3 448 73
NM_001272779.1 NP_001259708.1 6 - 7 2579 73
NM_134475.4 NP_608319.1 2 - 3 448 73
NM_167665.2 NP_728267.1 6 - 7 2034 73
NM_167666.3 NP_728268.1 6 - 7 2384 73
NM_167667.3 NP_728269.1 1 - 2 587 73
GenBank mRNA AB277547.1 - 6 - 7 2363 73
AB277548.1 - 2 - 3 374 73
AY069469.1 - 2 - 3 417 73
BT099719.1 - 6 - 7 2323 73

View Full Product Details