Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pyx
Entrez Gene ID
38037
Gene Name
pyrexia
Gene Aliases
CG17142, CT33412, Dmel\CG17142, Dmel_CG17142, Pyrexia, Pyx, dTRPA2, trpA2
Chromosome Location
Chr.3L: 217932 - 223069 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0035113
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138171.2 NP_612015.1 6 - 7 1623 96
NM_167813.2 NP_728501.1 2 - 3 159 96
GenBank mRNA BT001363.1 - 2 - 3 159 96
BT010317.1 - 6 - 7 1623 96

View Full Product Details