Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RhoGEF3
Entrez Gene ID
38050
Gene Name
Rho guanine nucleotide exchange factor 3
Gene Aliases
CG1225, CG13881, CG13882, CG13883, CG42377, CG42378, CG43976, Dmel\CG43976, Dmel_CG43976, DrhoGEF3, RhoGEF, rhoGEF3
Chromosome Location
Chr.3L: 277437 - 305292 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0264707
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144390.2 NP_001137862.1 6 - 7 1998 59
NM_001144391.1 NP_001137863.1 4 - 5 1911 59
NM_001169836.1 NP_001163307.1 6 - 7 1735 59
NM_001169837.1 NP_001163308.1 6 - 7 1810 59
NM_001274273.1 NP_001261202.1 15 - 16 9764 59
NM_001382119.1 NP_001368935.1 5 - 6 1862 59
NM_138180.4 NP_612024.4 15 - 16 9767 59
NM_138182.2 NP_612026.2 6 - 7 2154 59
NM_138183.3 NP_612027.2 6 - 7 1998 59
NM_167823.2 NP_728515.1 6 - 7 1827 59
NM_167824.2 NP_728516.1 6 - 7 1821 59
NM_167825.2 NP_728517.1 5 - 6 1806 59
GenBank mRNA BT011402.1 - 6 - 7 1827 59
BT016166.1 - 6 - 7 1780 59
BT044233.1 - 6 - 7 1978 59
BT099707.1 - 6 - 7 2154 59

View Full Product Details