Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sec22
Entrez Gene ID
31030
Gene Name
Secretory 22
Gene Aliases
34F3.8, CG7359, Dmel\CG7359, Dmel_CG7359, EG:34F3.8, SEC22, Sec22p
Chromosome Location
Chr.X: 806076 - 808200 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0260855
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130507.3 NP_569863.1 1 - 2 490 92
GenBank mRNA AY069680.1 - 1 - 2 460 92

View Full Product Details