Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mur2B
Entrez Gene ID
31111
Gene Name
Mucin related 2B
Gene Aliases
CG14796, Dmel\CG14796, Dmel_CG14796, EG:56G7.1
Chromosome Location
Chr.X: 1523554 - 1668788 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0025390
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001201595.2 NP_001188524.1 5 - 6 1283 94
NM_001297860.1 NP_001284789.1 6 - 7 1644 94
NM_130571.3 NP_569927.1 5 - 6 1283 94

View Full Product Details