Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Gr59d
- Entrez Gene ID
-
117343
- Gene Name
- Gustatory receptor 59d
- Gene Aliases
-
59D.1, 59D1, CG13543, CG30330, Dmel\CG30330, Dmel_CG30330, GR59D.1, GR59d, Gr59D1
- Chromosome Location
-
Chr.2R: 23103941 - 23105169 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0041236
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_137914.2 |
NP_611758.2 |
1 - 2 |
1013 |
65 |
View Full Product Details