Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
snama
Entrez Gene ID
37857
Gene Name
something that sticks like glue
Gene Aliases
BCDNA:LD21643, BcDNA:LD21643, CG3231, Dmel\CG3231, Dmel_CG3231, FBgn0086129, SNAMA, Snama, l(2)rQ313, mnm
Chromosome Location
Chr.2R: 24086604 - 24092014 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0086129
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001259558.2 NP_001246487.1 3 - 4 652 69
NM_138040.4 NP_611884.1 3 - 4 524 69
GenBank mRNA AF132177.1 - 3 - 4 496 69
BT124892.1 - 3 - 4 862 69

View Full Product Details