Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zfrp8
Entrez Gene ID
45021
Gene Name
Zinc finger protein RP-8
Gene Aliases
137/5, CG3260, Dmel\CG3260, Dmel_CG3260, PDCD2, l(2)k13705, zfrp8
Chromosome Location
Chr.2R: 24133989 - 24135405 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0021875
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001299899.1 NP_001286828.1 1 - 2 297 86
NM_138046.2 NP_611890.1 1 - 2 297 86

View Full Product Details