Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Ir60a
- Entrez Gene ID
-
37881
- Gene Name
- Ionotropic receptor 60a
- Gene Aliases
-
CG13576, DmelIR60a, Dmel\CG13576, Dmel_CG13576, IR60a
- Chromosome Location
-
Chr.2R: 24249055 - 24251303 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0034994
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_138057.1 |
NP_611901.1 |
1 - 2 |
916 |
113 |
View Full Product Details