Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- yellow-h
- Entrez Gene ID
-
43779
- Gene Name
- CG1629-PA
- Gene Aliases
-
CG1629, Dmel\CG1629, Dmel_CG1629, h, yell-h
- Chromosome Location
-
Chr.4: 227922 - 230428 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0039896
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_143655.4 |
NP_651912.3 |
2 - 3 |
986 |
86 |
View Full Product Details