Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lgs
Entrez Gene ID
43791
Gene Name
legless
Gene Aliases
BCL9, CG2041, Dm Lgs, Dmel\CG2041, Dmel_CG2041, Lgl, Lgs
Chromosome Location
Chr.4: 436957 - 443911 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0039907
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_143665.4 NP_651922.1 4 - 5 3204 86
GenBank mRNA AY051651.1 - 4 - 5 3133 86

View Full Product Details