Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Spt6
Entrez Gene ID
44000
Gene Name
CG12225-PA
Gene Aliases
CG12225, Dmel\CG12225, Dmel_CG12225, Dspt6, SPT6, dSpt6, dspt6, l(1)G0063, spt 6, spt6
Chromosome Location
Chr.X: 6267797 - 6274984 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0028982
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001298007.1 NP_001284936.1 3 - 4 443 75
NM_143705.3 NP_651962.2 3 - 4 443 75
GenBank mRNA BT150092.1 - 3 - 4 417 75
BT150473.1 - 3 - 4 417 75

View Full Product Details