Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ScsbetaG
Entrez Gene ID
44001
Gene Name
Succinyl-coenzyme A synthetase beta subunit, GDP-forming
Gene Aliases
142911_at, CG10622, Dmel\CG10622, Dmel_CG10622, G-Sbeta, SucB, Sucb, sucB, sucb
Chromosome Location
Chr.3L: 5561701 - 5563691 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0029118
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300012.1 NP_001286941.1 1 - 2 196 64
NM_143706.5 NP_651963.1 1 - 2 196 64
GenBank mRNA AJ252069.1 - 1 - 2 163 64
AY051980.1 - 1 - 2 148 64

View Full Product Details