Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cyp6a17
Entrez Gene ID
45556
Gene Name
Cytochrome P450-6a17
Gene Aliases
61D6S, 6a17, CG10241, Cyp6A17, Dmel\CG10241, Dmel_CG10241, ESTS:61D6S, cyp6a17, temp
Chromosome Location
Chr.2R: 14873953 - 14875739 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0015714
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001299504.1 NP_001286433.1 1 - 2 1115 103
NM_143761.4 NP_652018.1 1 - 2 1115 103
GenBank mRNA AY069090.1 - 1 - 2 1096 103

View Full Product Details