Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Spn27A
Entrez Gene ID
45815
Gene Name
Serpin 27A
Gene Aliases
CG11331, Dm-Spn27A, Dmel\CG11331, Dmel_CG11331, EST1, Spn27, Spn27a, sp27A, spn27A
Chromosome Location
Chr.2L: 6671065 - 6673347 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0028990
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273214.1 NP_001260143.1 1 - 1 1514 67
NM_143767.3 NP_652024.1 1 - 1 1514 67
GenBank mRNA AF220045.1 - 1 - 1 1487 67
BT004486.1 - 1 - 1 1486 67
BT125777.1 - 1 - 1 1486 67

View Full Product Details