Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Prosbeta1
- Entrez Gene ID
-
46058
- Gene Name
- Proteasome beta1 subunit
- Gene Aliases
-
1(2)05070, 8392, B[[1]], Beta-1, CG8392, Dmel\CG8392, Dmel_CG8392, beta-1, beta1, l(2)05070, prosbeta1
- Chromosome Location
-
Chr.2R: 16013805 - 16014787 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0010590
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_143774.3 |
NP_652031.2 |
1 - 2 |
139 |
74 |
GenBank mRNA |
BT044319.1 |
- |
1 - 2 |
133 |
74 |
View Full Product Details