Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hipk
Entrez Gene ID
38070
Gene Name
Homeodomain interacting protein kinase
Gene Aliases
CG17090, DHIPK2, Dmel\CG17090, Dmel_CG17090, HIPK, HIPK2, dhipk2, hipK, hipk
Chromosome Location
Chr.3L: 543060 - 581099 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0035142
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274286.1 NP_001261215.1 7 - 8 2784 126
NM_001382035.1 NP_001369036.1 8 - 9 3463 126
NM_138194.3 NP_612038.2 8 - 9 3385 126
NM_167834.2 NP_728531.1 8 - 9 3463 126

View Full Product Details