Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ptp61F
Entrez Gene ID
38160
Gene Name
Protein tyrosine phosphatase 61F
Gene Aliases
BEST:LP01280, CG9178, CG9181, CPtp62A, DCPTP62A, DPTP61F, Dmel\CG9181, Dmel_CG9181, PTP1B, PTP61F, Ptp-61F, Ptp61f, R-PTP 61F, anon-WO0118547.297, anon-WO0140519.98, dPTP61F, dptp61F, ptp61F
Chromosome Location
Chr.3L: 1342493 - 1475257 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0267487
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_057340.4 NP_476688.1 6 - 7 1696 67
NM_167874.2 NP_728600.1 6 - 7 1657 67
GenBank mRNA AY069713.1 - 6 - 7 1657 67

View Full Product Details