Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Psa
- Entrez Gene ID
-
38175
- Gene Name
- Puromycin sensitive aminopeptidase
- Gene Aliases
-
CG1009, Dmel\CG1009, Dmel_CG1009, PSA, dPSA, dPsa, l(3)06226
- Chromosome Location
-
Chr.3L: 1502843 - 1523380 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0261243
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_167883.2 |
NP_728614.1 |
2 - 3 |
218 |
74 |
GenBank mRNA |
BT133253.1 |
- |
2 - 3 |
172 |
74 |
View Full Product Details