Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Iml1
Entrez Gene ID
38176
Gene Name
Increased minichromosome loss 1
Gene Aliases
CG12090, Dmel\CG12090, Dmel_CG12090, SEA1, iml1
Chromosome Location
Chr.3L: 1523414 - 1534034 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0035227
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274330.1 NP_001261259.1 15 - 16 3659 69
NM_001274331.1 NP_001261260.1 15 - 16 3635 69
NM_139361.2 NP_647618.1 14 - 15 3539 69
NM_167888.2 NP_728620.1 14 - 15 3542 69
NM_167889.2 NP_728621.1 16 - 17 3758 69

View Full Product Details