Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GV1
Entrez Gene ID
38238
Gene Name
CG12023-PA
Gene Aliases
BcDNA:RH49123, CG12023, DIP1, Dmel\CG12023, Dmel_CG12023
Chromosome Location
Chr.3L: 1810588 - 1822903 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0027790
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139408.3 NP_647665.2 3 - 4 398 69
NM_167920.2 NP_728678.1 3 - 4 398 69
GenBank mRNA AY075547.1 - 3 - 4 397 69

View Full Product Details