Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dre4
Entrez Gene ID
38248
Gene Name
CG1828-PA
Gene Aliases
CG1828, DRE4, DRE4/dSPT16, Dmel\CG1828, Dmel_CG1828, Dre4, SPT16, Spt16, dSPT16, dSpt16, dspt16, l(3)62Bg, l(3)dre4, spt16
Chromosome Location
Chr.3L: 1871574 - 1876312 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0002183
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_057262.4 NP_476610.2 4 - 5 1951 66
NM_167922.3 NP_728686.2 4 - 5 1951 66
GenBank mRNA BT010116.1 - 4 - 5 1975 66

View Full Product Details