Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
promL
Entrez Gene ID
38372
Gene Name
prominin-like
Gene Aliases
CG7740, Dmel\CG7740, Dmel_CG7740, bs29e02.y1
Chromosome Location
Chr.3L: 3114450 - 3127724 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0026189
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001259658.3 NP_001246587.1 13 - 14 3231 101
NM_001274422.1 NP_001261351.1 13 - 14 3208 101
NM_001274424.1 NP_001261353.1 12 - 13 3350 101
NM_139513.3 NP_647770.1 13 - 14 3132 101
NM_167986.1 NP_728809.1 13 - 14 3410 101
NM_167987.4 NP_728810.1 14 - 15 3291 101
GenBank mRNA AF127935.2 - 14 - 15 3266 101
AF197345.1 - 13 - 14 3088 101
BT031128.1 - 13 - 14 3090 101

View Full Product Details